Anti-PRKG2 Rabbit Polyclonal Antibody
Catalog # 77723-242
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:PRKG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Form:liquid
- Gene ID:Q13237
- Antigen synonyms:cGMP-dependent protein kinase II|PRKG 2|KGP2_HUMAN|PRKG2|cGMP dependent|type II|CGK 2|PRKGR2|Type II cGMP dependent protein kinase|cGK2|Protein kinase|cGMP-dependent protein kinase 2|cGKII|cGMP dependent protein kinase 2
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Synthesized non-phosphopeptide derived from Human PKG2 around the phosphorylation site of serine 126 (G-V-S(p)-A-E).
- Purification:Antigen affinity purified
- Cat. no.:77723-242
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: PRKG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse