Anti-AADAT Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: AADAT
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77723-043
- 77723-044
- Antigen symbol:
- AADAT
- AADAT
- Antigen synonyms:
- L kynurenine/alpha aminoadipate aminotransferase|Aminoadipate aminotransferase|2 aminoadipate transaminase|Alpha-aminoadipate aminotransferase|KATII |Kynurenine oxoglutarate transaminase 2|Aadt|Kynurenine oxoglutarate transaminase II|Aadat|Kynurenine aminotransferase II|AI875679|mitochondrial|2-aminoadipate transaminase|AADAT_HUMAN|2-aminoadipate aminotransferase|Kynurenine/alpha-aminoadipate aminotransferase|Kynurenine--oxoglutarate aminotransferase II|KAT2|Kynurenine oxoglutarate aminotransferase II|EC 2.6.1.7|EC 2.6.1.39|2 aminoadipate aminotransferase|Kynurenine--oxoglutarate transaminase II|Alpha aminoadipate aminotransferase|Kynurenine/alpha-aminoadipate aminotransferase mitochondrial [Precursor]|KAT/AadAT
- L kynurenine/alpha aminoadipate aminotransferase|Aminoadipate aminotransferase|2 aminoadipate transaminase|Alpha-aminoadipate aminotransferase|KATII |Kynurenine oxoglutarate transaminase 2|Aadt|Kynurenine oxoglutarate transaminase II|Aadat|Kynurenine aminotransferase II|AI875679|mitochondrial|2-aminoadipate transaminase|AADAT_HUMAN|2-aminoadipate aminotransferase|Kynurenine/alpha-aminoadipate aminotransferase|Kynurenine--oxoglutarate aminotransferase II|KAT2|Kynurenine oxoglutarate aminotransferase II|EC 2.6.1.7|EC 2.6.1.39|2 aminoadipate aminotransferase|Kynurenine--oxoglutarate transaminase II|Alpha aminoadipate aminotransferase|Kynurenine/alpha-aminoadipate aminotransferase mitochondrial [Precursor]|KAT/AadAT
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N5Z0
- Q8N5Z0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kynurenine/alpha-aminoadipate aminotransferase, mitochondrial protein (30-425AA)
- Recombinant Human Kynurenine/alpha-aminoadipate aminotransferase, mitochondrial protein (30-425AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together