Anti-RDH12 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RDH12
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77722-835
- 77722-836
- Antigen symbol:
- RDH12
- RDH12
- Antigen synonyms:
- Retinol dehydrogenase 12|All trans and 9 cis retinol dehydrogenase|RDH 12|LCA13|LCA3|RDH12|RP53|LCA 3|All-trans and 9-cis retinol dehydrogenase|Retinol dehydrogenase 12 (all trans/9 cis/11 cis)|RDH12_HUMAN|Retinol dehydrogenase 12 all trans and 9 cis|Short chain dehydrogenase/reductase family 7C member 2|SDR7C2
- Retinol dehydrogenase 12|All trans and 9 cis retinol dehydrogenase|RDH 12|LCA13|LCA3|RDH12|RP53|LCA 3|All-trans and 9-cis retinol dehydrogenase|Retinol dehydrogenase 12 (all trans/9 cis/11 cis)|RDH12_HUMAN|Retinol dehydrogenase 12 all trans and 9 cis|Short chain dehydrogenase/reductase family 7C member 2|SDR7C2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96NR8
- Q96NR8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Retinol dehydrogenase 12 protein (1-316AA)
- Recombinant Human Retinol dehydrogenase 12 protein (1-316AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together