Order Entry
Canada
ContactUsLinkComponent
Anti-CDS1 Rabbit Polyclonal Antibody (HRP)
Anti-CDS1 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
Anti-CDS1 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77722-475
  • 77722-476
  • Antigen symbol:
  • CDS1
  • CDS1
  • Antigen synonyms:
  • CDP DG synthetase 1|CDP diacylglycerol synthase 1|CDP-diglyceride pyrophosphorylase 1|CDS1|CDP diacylglycerol synthase (phosphatidate cytidylyltransferase) 1|CDP-diacylglycerol synthase 1|CDP-DG synthase 1|CDS 1|CDP-DAG synthase 1|CDP diglyceride pyrophosphorylase 1|CDP DAG synthase 1|CTP:phosphatidate cytidylyltransferase 1|CDS|CDP diglyceride synthetase 1|Phosphatidate cytidylyltransferase 1|CDP-diglyceride synthase 1|CDS1_HUMAN
  • CDP DG synthetase 1|CDP diacylglycerol synthase 1|CDP-diglyceride pyrophosphorylase 1|CDS1|CDP diacylglycerol synthase (phosphatidate cytidylyltransferase) 1|CDP-diacylglycerol synthase 1|CDP-DG synthase 1|CDS 1|CDP-DAG synthase 1|CDP diglyceride pyrophosphorylase 1|CDP DAG synthase 1|CTP:phosphatidate cytidylyltransferase 1|CDS|CDP diglyceride synthetase 1|Phosphatidate cytidylyltransferase 1|CDP-diglyceride synthase 1|CDS1_HUMAN
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q92903
  • Q92903
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Phosphatidate cytidylyltransferase 1 protein (6-85AA)
  • Recombinant Human Phosphatidate cytidylyltransferase 1 protein (6-85AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together