Anti-RELA (phospho–Ser529) Rabbit Polyclonal Antibody
77716-309
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen symbol:RELA (phospho-Ser529)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Form:liquid
- Gene ID:Q04206
- Antigen synonyms:OTTHUMP00000233474|TF65_HUMAN|p65 NFkB|V rel avian reticuloendotheliosis viral oncogene homolog A|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3|v rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor of kappa light polypeptide gene enhancer in B cells 3 (p65))|Nuclear Factor NF Kappa B p65 Subunit|p65|v rel reticuloendotheliosis viral oncogene homolog A (avian)|p65 NF kappaB|Transcription factor NFKB3|Nuclear factor kappaB|V rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappa light polypeptide gene enhancer in B cells 3, p65|OTTHUMP00000233473|MGC131774|OTTHUMP00000233476|NF kappa B p65delta3|nfkappabp65|Transcription factor p65|NFKB3|Nuclear factor of kappa light polypeptide gene enhancer in B cells 3|relA|NFkB p65|OTTHUMP00000233900|Avian reticuloendotheliosis viral (v rel) oncogene homolog A|Nuclear factor NF-kappa-B p65 subunit|OTTHUMP00000233475
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Peptide sequence around phosphorylation site of serine 529 (L-L-S(p)-G-D) derived from Human NFkB-p65.
- Purification:Antigen affinity purified
- Cat. no.:77716-309
- Packaging:Vial
WB:1:500-1:1000, IHC:1:50-1:100, IF:1:100-1:200
Type:
Antigen: RELA (phospho–Ser529)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat