Anti-RUNX1T1 Rabbit Polyclonal Antibody
77689-788
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen symbol:RUNX1T1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q06455
- Antigen synonyms:ETO protein|MTG 8|Core binding factor runt domain alpha subunit 2 translocated to 1 cyclin D related|Cyclin-D-related protein|CBFA2T1|ZMYND2|AML1 MTG8|Runt related transcription factor 1 translocated to 1 (cyclin D related)|Runt related transcription factor 1 translocated to 1 cyclin D related|Eight twenty one protein|Protein ETO|MTG8 protein|AML1T1|Acute myelogenous leukemia 1 translocation 1 protein|Protein MTG 8|Myeloid translocation gene on 8q22|Runt related transcription factor 1 translocated to 1|Core binding factor runt domain alpha subunit 2 translocated to 1|Cyclin D related protein|CDR|MTG8_HUMAN|ZMYND 2|RUNX1T1|MTG8|Zinc finger MYND domain-containing protein 2|Protein CBFA2T1|MGC2796|RUNX1 translocation partner 1|MTG 8b|Acute myelogenous leukemia 1 translocation 1 cyclin D related|Protein MTG8|MTG8b|Cyclin D related|Zinc finger MYND domain containing protein 2|ETO
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Protein CBFA2T1 protein (416-525AA)
- Purification:>95%, Protein G purified
- Cat. no.:77689-788
- Packaging:Vial
Recommended dilution: WB:1:2000-1:10000, IHC:1:20-1:200
Type:
Antigen: RUNX1T1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human