Anti-AGK Rabbit Polyclonal Antibody
77689-737
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen symbol:AGK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q53H12
- Antigen synonyms:AGK_HUMAN|Acylglycerol kinase mitochondrial|6720408I04Rik|Multi substrate lipid kinase|mitochondrial|Acylglycerol kinase|Acylglycerol Kinase|agk|FLJ10842|Multiple substrate lipid kinase|Multi-substrate lipid kinase|2610037M15Rik|AI465370|MuLK|RGD1562046|HsMuLK|hAGK
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Acylglycerol kinase, mitochondrial protein (32-65AA)
- Purification:>95%, Protein G purified
- Cat. no.:77689-737
- Packaging:Vial
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: AGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse