Anti-KIF23 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200-1:500
Type:
Antigen: KIF23
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77714-736
- 77714-737
- Antigen symbol:
- KIF23
- KIF23
- Antigen synonyms:
- CHO 1|Kinesin like protein KIF23|KIF 23|Kinesin family member 23|KIF23|KNS L5|CHO1|Kinesin like 5|KIF23_HUMAN|KNSL 5|Mitotic kinesin like protein 1|KNSL5|Kinesin like protein KIF 23|Kinesin like protein 5|MKLP 1|Mitotic kinesin like 1|Kinesin-like protein KIF23|Kinesin-like protein 5|Mitotic kinesin-like protein 1
- CHO 1|Kinesin like protein KIF23|KIF 23|Kinesin family member 23|KIF23|KNS L5|CHO1|Kinesin like 5|KIF23_HUMAN|KNSL 5|Mitotic kinesin like protein 1|KNSL5|Kinesin like protein KIF 23|Kinesin like protein 5|MKLP 1|Mitotic kinesin like 1|Kinesin-like protein KIF23|Kinesin-like protein 5|Mitotic kinesin-like protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02241
- Q02241
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kinesin-like protein KIF23 protein (865-960AA)
- Recombinant Human Kinesin-like protein KIF23 protein (865-960AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together