Order Entry
Canada
ContactUsLinkComponent
 
Anti-UGT1A10 Rabbit Polyclonal Antibody
New Product
 
Anti-UGT1A10 Rabbit Polyclonal Antibody
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 77714-271
  • 77714-270
  • Antigen symbol:
  • UGT1A10
  • UGT1A10
  • Antigen synonyms:
  • UDP-glucuronosyltransferase 1A10|UDPGT 1-10|GNT1|UGT1UDP-glucuronosyltransferase 1-10|UGT-1J|UGT1.10|UGT1-10|UGT1J|UGT1*10|UDP-glucuronosyltransferase 1-J|EC 2.4.1.17|UGT1A10
  • UDP-glucuronosyltransferase 1A10|UDPGT 1-10|GNT1|UGT1UDP-glucuronosyltransferase 1-10|UGT-1J|UGT1.10|UGT1-10|UGT1J|UGT1*10|UDP-glucuronosyltransferase 1-J|EC 2.4.1.17|UGT1A10
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9HAW8
  • Q9HAW8
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human UGT1A10
  • Fusion protein of Human UGT1A10
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together