Anti-ERI1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERI1
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77714-049
- 77714-048
- Antigen symbol:
- ERI1
- ERI1
- Antigen synonyms:
- Histone mRNA 3''-exonuclease 1|Eri 1 homolog|3' 5' exonuclease ERI 1|Three prime histone mRNA exonuclease 1|eri1|3' EXO|Histone mRNA 3' end specific exoribonuclease|Eri-1 homolog|Histone mRNA 3' end specific exonuclease|3'EXO|ERI 1|3'HEXO|3' 5' exonuclease ERI1|3''-5'' exonuclease eri1|3' HEXO|ERI1_HUMAN|THEX 1|Eri1 homolog|3''-5'' exoribonuclease 1|MGC35395|HEXO|THEX1|Protein 3''hExo|Histone mRNA 3''-end-specific exoribonuclease|3' exoribonuclease|3' histone mRNA exonuclease 1
- Histone mRNA 3''-exonuclease 1|Eri 1 homolog|3' 5' exonuclease ERI 1|Three prime histone mRNA exonuclease 1|eri1|3' EXO|Histone mRNA 3' end specific exoribonuclease|Eri-1 homolog|Histone mRNA 3' end specific exonuclease|3'EXO|ERI 1|3'HEXO|3' 5' exonuclease ERI1|3''-5'' exonuclease eri1|3' HEXO|ERI1_HUMAN|THEX 1|Eri1 homolog|3''-5'' exoribonuclease 1|MGC35395|HEXO|THEX1|Protein 3''hExo|Histone mRNA 3''-end-specific exoribonuclease|3' exoribonuclease|3' histone mRNA exonuclease 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IV48
- Q8IV48
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 3\'-5\' exoribonuclease 1 protein (2-66AA)
- Recombinant Human 3\'-5\' exoribonuclease 1 protein (2-66AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together