Anti-CEBPD Rabbit Polyclonal Antibody
New Product
ELISA:1:500-1:5000, WB:1:200-1:1000
Type:
Antigen: CEBPD
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77713-373
- 77713-374
- Antigen synonyms:
- Crp 3|CCAAT/enhancer binding protein (C/EBP) delta|CEBPD|CEBP D|CCAAT/enhancer binding protein delta|Nuclear factor NF IL6 beta|Nuclear factor NF-IL6-beta|NF-IL6-beta|C/EBP delta|C/EBP related protein 3|Crp3|NF IL6 beta|CEBPD_HUMAN|CELF|CCAAT/enhancer-binding protein delta
- Crp 3|CCAAT/enhancer binding protein (C/EBP) delta|CEBPD|CEBP D|CCAAT/enhancer binding protein delta|Nuclear factor NF IL6 beta|Nuclear factor NF-IL6-beta|NF-IL6-beta|C/EBP delta|C/EBP related protein 3|Crp3|NF IL6 beta|CEBPD_HUMAN|CELF|CCAAT/enhancer-binding protein delta
- Gene ID:
- P49716
- P49716
- Immunogen:
- Synthetic peptide of Human CEBPD
- Synthetic peptide of Human CEBPD
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together