Anti-THAP12 Rabbit Polyclonal Antibody
Catalog # 77688-497
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:THAP12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O43422
- Antigen synonyms:58 kDa interferon-induced protein kinase-interacting protein|P52RIPK|52 kDa repressor of the inhibitor of the protein kinase|p52rIPK|THAP domain-containing protein 12|PRKRIR|p58IPK-interacting protein|THAP domain-containing protein 0|Death-associated protein 4|DAP4|THAP0|THAP12
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human 52 kDa repressor of the inhibitor of the protein kinase protein (612-761AA)
- Purification:>95%, Protein G purified
- Cat. no.:77688-497
- Packaging:Vial
Specifications
About this item
Type:
Antigen: THAP12
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human