Anti-PILRB Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:20-1:100
Type:
Antigen: PILRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77712-204
- 77712-203
- Antigen symbol:
- PILRB
- PILRB
- Antigen synonyms:
- Paired immunoglobulin like receptor beta|Activating receptor PILRbeta|FDFACT|FDFACT1|Pilrb|Activating receptor PILR beta|Cell surface receptor FDFACT|Paired immunoglobulin like type 2 receptor beta|Cell surface receptor FDFACT2|Cell surface receptor FDFACT1|PILRB_HUMAN|Paired immunoglobin like receptor beta|Activating receptor PILR-beta|Paired immunoglobulin like type 2 receptor beta precursor|Paired immunoglobulin-like type 2 receptor beta|FDFACT2
- Paired immunoglobulin like receptor beta|Activating receptor PILRbeta|FDFACT|FDFACT1|Pilrb|Activating receptor PILR beta|Cell surface receptor FDFACT|Paired immunoglobulin like type 2 receptor beta|Cell surface receptor FDFACT2|Cell surface receptor FDFACT1|PILRB_HUMAN|Paired immunoglobin like receptor beta|Activating receptor PILR-beta|Paired immunoglobulin like type 2 receptor beta precursor|Paired immunoglobulin-like type 2 receptor beta|FDFACT2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UKJ0
- Q9UKJ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PILRB
- Fusion protein of Human PILRB
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together