Anti-GPS1 Rabbit Polyclonal Antibody
Catalog # 77702-680
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:GPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q13098
- Antigen synonyms:COPS1|Signalosome subunit 1|CSN 1|Gps1|FUS6/COP11|Protein GPS1|Arabidopsis|GPS-1|CSN1_HUMAN|JAB1-containing signalosome subunit 1|SGN1|G protein pathway suppressor 1|COP9 signalosome complex subunit 1|homolog of|COP9 signalosome subunit 1|Protein MFH
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human CSN1 around the non-phosphorylation site of S454.
- Purification:Antigen affinity purified
- Cat. no.:77702-680
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: GPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat