Anti-AGPS Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: AGPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77711-412
- 77711-413
- Antigen symbol:
- AGPS
- AGPS
- Antigen synonyms:
- AGPS|Alkyldihydroxyacetonephosphate synthase, peroxisomal|Aging associated gene 5 protein|Aging-associated gene 5 protein|ADAP-S|peroxisomal|Alkyldihydroxyacetonephosphate synthase|ALDHPSY|Alkylglycerone-phosphate synthase|Alkylglycerone phosphate synthase|ADAS|RCDP3|ADHAPS|AAG5|Alkyl-DHAP synthase|aging-associated protein 5|ADPS|ADAS_HUMAN
- AGPS|Alkyldihydroxyacetonephosphate synthase, peroxisomal|Aging associated gene 5 protein|Aging-associated gene 5 protein|ADAP-S|peroxisomal|Alkyldihydroxyacetonephosphate synthase|ALDHPSY|Alkylglycerone-phosphate synthase|Alkylglycerone phosphate synthase|ADAS|RCDP3|ADHAPS|AAG5|Alkyl-DHAP synthase|aging-associated protein 5|ADPS|ADAS_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00116
- O00116
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human AGPS
- Fusion protein of Human AGPS
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together