Anti-TNFRSF13C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: TNFRSF13C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77715-462
- 77715-461
- Antigen symbol:
- TNFRSF13C
- TNFRSF13C
- Antigen synonyms:
- BR 3|Tumor necrosis factor receptor subunit member 13C|CVID4|B cell activating factor receptor|BAFFR|CD 268|BAFF R|BR3|Prolixin|BAFF-R|BLyS receptor 3|Tnfrsf13c|BROMIX|CD268|CD268 antigen|TR13C_HUMAN|B-cell-activating factor receptor|Tumor necrosis factor receptor superfamily member 13C|TNFRSF 13C|BlySR3|MGC138235|OTTHUMP00000028746|BAFF receptor
- BR 3|Tumor necrosis factor receptor subunit member 13C|CVID4|B cell activating factor receptor|BAFFR|CD 268|BAFF R|BR3|Prolixin|BAFF-R|BLyS receptor 3|Tnfrsf13c|BROMIX|CD268|CD268 antigen|TR13C_HUMAN|B-cell-activating factor receptor|Tumor necrosis factor receptor superfamily member 13C|TNFRSF 13C|BlySR3|MGC138235|OTTHUMP00000028746|BAFF receptor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RJ3
- Q96RJ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TNFRSF13C
- Synthetic peptide of Human TNFRSF13C
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together