Anti-GOLPH3 Rabbit Polyclonal Antibody
Catalog # 77690-751
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:GOLPH3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9H4A6
- Antigen synonyms:GPP34|GOPP1|Golgi phosphoprotein 3|Golgi phosphoprotein 3 (coat protein)|Golgi associated protein|GOLP3_HUMAN|Mitochondrial DNA absence factor|MIDAS|FLJ90675|Golph3|Coat protein GPP34|Golgi protein|Golgi peripheral membrane protein 1, 34 kDa|Coat protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Golgi phosphoprotein 3 protein (16-150AA)
- Purification:>95%, Protein G purified
- Cat. no.:77690-751
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:20-1:200, IP:1:200-1:2000
Type:
Antigen: GOLPH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human