Anti-LILRB1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:200-1:500, IF:1:50-1:200
Type:
Antigen: LILRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77690-389
- 77724-326
- Antigen symbol:
- LILRB1
- LILRB1
- Antigen synonyms:
- Ig like transcript 2|PIRB|ILT-2|LIR-1|immunoglobulin like transcript 2|CD85j antigen|LIR1|leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1|CD85|LIR 1|Leukocyte immunoglobulin-like receptor 1|CD85 antigen|leukocyte immunoglobulin like receptor, subfamily B, member 1|MIR7|MIR-7|LILRB1|CD85 antigen like family member J|Leukocyte immunoglobulin like receptor 1|MIR 7|Monocyte/macrophage immunoglobulin like receptor 7|leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform|leukocyte immunoglobulin like receptor subfamily B member 1 soluble isoform|Leukocyte immunoglobulin-like receptor subfamily B member 1|ILT2|leukocyte Ig like receptor 1|Immunoglobulin-like transcript 2|leukocyte immunoglobulin like receptor subfamily B member 1|CD85j|PIR B|ILT 2|Monocyte/macrophage immunoglobulin-like receptor 7|CD85 antigen-like family member J|Leukocyte immunoglobulin like receptor subfamily B member 1 precursor|LIRB1_HUMAN
- Ig like transcript 2|PIRB|ILT-2|LIR-1|immunoglobulin like transcript 2|CD85j antigen|LIR1|leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1|CD85|LIR 1|Leukocyte immunoglobulin-like receptor 1|CD85 antigen|leukocyte immunoglobulin like receptor, subfamily B, member 1|MIR7|MIR-7|LILRB1|CD85 antigen like family member J|Leukocyte immunoglobulin like receptor 1|MIR 7|Monocyte/macrophage immunoglobulin like receptor 7|leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform|leukocyte immunoglobulin like receptor subfamily B member 1 soluble isoform|Leukocyte immunoglobulin-like receptor subfamily B member 1|ILT2|leukocyte Ig like receptor 1|Immunoglobulin-like transcript 2|leukocyte immunoglobulin like receptor subfamily B member 1|CD85j|PIR B|ILT 2|Monocyte/macrophage immunoglobulin-like receptor 7|CD85 antigen-like family member J|Leukocyte immunoglobulin like receptor subfamily B member 1 precursor|LIRB1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NHL6
- Q8NHL6
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leukocyte immunoglobulin-like receptor subfamily B member 1 protein (483-650AA)
- Recombinant Human Leukocyte immunoglobulin-like receptor subfamily B member 1 protein (483-650AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together