Anti-DCP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: DCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77717-265
- 77717-264
- Antigen symbol:
- DCP2
- DCP2
- Antigen synonyms:
- DCP2, S. cerevisiae, homolog of|Nudix motif 20|mRNA decapping enzyme 2|FLJ33245|decapping enzyme 2, S. cerevisiae, homolog of|decapping enzyme homolog (S. cerevisiae)|Nucleoside diphosphate-linked moiety X motif 20|NUDT20|nudix (nucleoside diphosphate linked moiety X)-type motif 20|m7GpppN-mRNA hydrolase|mRNA-decapping enzyme 2|DCP2|DCP2_HUMAN|DCP2 decapping enzyme homolog (S. cerevisiae)|hDpc
- DCP2, S. cerevisiae, homolog of|Nudix motif 20|mRNA decapping enzyme 2|FLJ33245|decapping enzyme 2, S. cerevisiae, homolog of|decapping enzyme homolog (S. cerevisiae)|Nucleoside diphosphate-linked moiety X motif 20|NUDT20|nudix (nucleoside diphosphate linked moiety X)-type motif 20|m7GpppN-mRNA hydrolase|mRNA-decapping enzyme 2|DCP2|DCP2_HUMAN|DCP2 decapping enzyme homolog (S. cerevisiae)|hDpc
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IU60
- Q8IU60
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human m7GpppN-mRNA hydrolase protein (1-385AA)
- Recombinant Human m7GpppN-mRNA hydrolase protein (1-385AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together