Anti-DND1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: DND1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77717-203
- 77717-202
- Antigen symbol:
- DND1
- DND1
- Antigen synonyms:
- DND microRNA-mediated repression inhibitor 1|homolog|MGC34750|Ter|zebrafish|RNA binding motif single stranded interacting protein 4|DND1_HUMAN|dnd1|Dead end protein homolog 1|Dead end homolog 1 (zebrafish)|dead end homolog 1|Dead end protein|RBMS4|1|Dead end|single-stranded-interacting protein 4|RNA-binding motif
- DND microRNA-mediated repression inhibitor 1|homolog|MGC34750|Ter|zebrafish|RNA binding motif single stranded interacting protein 4|DND1_HUMAN|dnd1|Dead end protein homolog 1|Dead end homolog 1 (zebrafish)|dead end homolog 1|Dead end protein|RBMS4|1|Dead end|single-stranded-interacting protein 4|RNA-binding motif
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IYX4
- Q8IYX4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dead end protein homolog 1 protein (37-180AA)
- Recombinant Human Dead end protein homolog 1 protein (37-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together