Anti-CDC16 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: CDC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77707-897
- 77707-896
- Antigen symbol:
- CDC16
- CDC16
- Antigen synonyms:
- CDC16 protein|CDC16 homolog|CDC16_HUMAN|CDC 16|Anaphase-promoting complex subunit 6|Cyclosome subunit 6|CDC16 (cell division cycle 16 S. cerevisiae homolog)|Cell division cycle protein 16 homolog|CDC16Hs|Cell division cycle 16|Anaphase promoting complex subunit 6|Apc 6|ANAPC6|Cell division cycle 16 homolog|CUT9|APC6|Cdc16
- CDC16 protein|CDC16 homolog|CDC16_HUMAN|CDC 16|Anaphase-promoting complex subunit 6|Cyclosome subunit 6|CDC16 (cell division cycle 16 S. cerevisiae homolog)|Cell division cycle protein 16 homolog|CDC16Hs|Cell division cycle 16|Anaphase promoting complex subunit 6|Apc 6|ANAPC6|Cell division cycle 16 homolog|CUT9|APC6|Cdc16
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13042
- Q13042
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CDC16
- Synthetic peptide of Human CDC16
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice.
- Shipped on blue ice.
Specifications
Frequently Bought Together