Anti-DHX58 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: DHX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77719-060
- 77719-059
- Antigen symbol:
- DHX58
- DHX58
- Antigen synonyms:
- Protein D11Lgp2 homolog|LGP 2|Probable ATP dependent helicase LGP2|D11LGP2|DHX 58|DHX58|D11lgp2e|DEXH box polypeptide 58|RIG-I-like receptor LGP2|Probable ATP-dependent RNA helicase DHX58|LGP2|RLR|DEXH (Asp Glu X His) box polypeptide 58|RNA helicase LGP2|Probable ATP-dependent helicase LGP2|Probable ATP dependent RNA helicase DHX58|DHX58_HUMAN|Ortholog of mouse D11lgp2
- Protein D11Lgp2 homolog|LGP 2|Probable ATP dependent helicase LGP2|D11LGP2|DHX 58|DHX58|D11lgp2e|DEXH box polypeptide 58|RIG-I-like receptor LGP2|Probable ATP-dependent RNA helicase DHX58|LGP2|RLR|DEXH (Asp Glu X His) box polypeptide 58|RNA helicase LGP2|Probable ATP-dependent helicase LGP2|Probable ATP dependent RNA helicase DHX58|DHX58_HUMAN|Ortholog of mouse D11lgp2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96C10
- Q96C10
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Probable ATP-dependent RNA helicase DHX58 protein (389-678AA)
- Recombinant Human Probable ATP-dependent RNA helicase DHX58 protein (389-678AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together