Anti-DLC1 Rabbit Polyclonal Antibody
Catalog # 77708-615
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:DLC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q96QB1
- Antigen synonyms:START domain-containing protein 12|STARD 12|StAR-related lipid transfer protein 12|FLJ21120|Rho GTPase activating protein 7|DLC1|Rho-type GTPase-activating protein 7|StAR related lipid transfer (START) domain containing 12|ARHGAP 7|Dlc-1|HP protein|DLC 1|p122 RhoGAP|RHG07_HUMAN|Deleted in liver cancer 1 protein|Rho GTPase-activating protein 7|ARHGAP7|StARD12|StAR related lipid transfer protein 12|START domain containing protein 12|Rho type GTPase activating protein 7|KIAA1723|DLC1 Rho GTPase activating protein|HP|Deleted in liver cancer 1
- Storage buffer:PBS, 150bmM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human RHG07
- Purification:Antigen affinity purified
- Cat. no.:77708-615
- Packaging:Vial
Specifications
About this item
IHC:1:50-1:100, IF:1:100-1:500
Type:
Antigen: DLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse