Anti-MED14 Rabbit Polyclonal Antibody
Catalog # 77705-032
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:MED14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O60244
- Antigen synonyms:DRIP150|Transcriptional coactivator CRSP150|MED14_HUMAN|MED14|RGR1|Thyroid hormone receptor-associated protein complex 170 kDa component|RGR1 homolog|Mediator of RNA polymerase II transcription subunit 14|Trap170|Vitamin D3 receptor-interacting protein complex 150 kDa component|hRGR1|CRSP complex subunit 2|EXLM1|Activator-recruited cofactor 150 kDa component|Mediator complex subunit 14|ARC150|Cofactor required for Sp1 transcriptional activation subunit 2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CRSP150
- Purification:Antigen affinity purified
- Cat. no.:77705-032
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: MED14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse