Anti-U2AF1L4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: U2AF1L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77721-320
- 77721-319
- Antigen symbol:
- U2AF1L4
- U2AF1L4
- Antigen synonyms:
- U2 small nuclear RNA auxiliary factor 1 like 4|U2(RNU2) small nuclear RNA auxiliary factor 1 like 3|U2 small nuclear RNA auxiliary factor 1 like protein 4|U2AF1L3V1|MGC33901|U2RNU2 small nuclear RNA auxiliary factor 1 like 3|U2 small nuclear RNA auxiliary factor 1 like 3|U2AF1RS3|U2RNU2 small nuclear RNA auxiliary factor 1 like protein 3|U2AF1-like protein 3|U2af26|U2AF1L4|U2AF4_HUMAN|Splicing factor U2AF 26 kDa subunit|FLJ35525|U2 small nuclear RNA auxiliary factor 1-like protein 4|U2 auxiliary factor 26|U2 small nuclear RNA auxiliary factor 1 like protein 3|U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3|U2AF1 RS3|U2AF1 like protein 3|U2 small nuclear RNA auxiliary factor 1-like protein 3
- U2 small nuclear RNA auxiliary factor 1 like 4|U2(RNU2) small nuclear RNA auxiliary factor 1 like 3|U2 small nuclear RNA auxiliary factor 1 like protein 4|U2AF1L3V1|MGC33901|U2RNU2 small nuclear RNA auxiliary factor 1 like 3|U2 small nuclear RNA auxiliary factor 1 like 3|U2AF1RS3|U2RNU2 small nuclear RNA auxiliary factor 1 like protein 3|U2AF1-like protein 3|U2af26|U2AF1L4|U2AF4_HUMAN|Splicing factor U2AF 26 kDa subunit|FLJ35525|U2 small nuclear RNA auxiliary factor 1-like protein 4|U2 auxiliary factor 26|U2 small nuclear RNA auxiliary factor 1 like protein 3|U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3|U2AF1 RS3|U2AF1 like protein 3|U2 small nuclear RNA auxiliary factor 1-like protein 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU68
- Q8WU68
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Splicing factor U2AF 26 kDa subunit protein (1-202AA)
- Recombinant Human Splicing factor U2AF 26 kDa subunit protein (1-202AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together