Anti-SUV39H2 Rabbit Polyclonal Antibody
Catalog # 77699-837
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:SUV39H2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9H5I1
- Antigen synonyms:Histone-lysine N-methyltransferase SUV39H2|FLJ23414|Lysine N methyltransferase 1B|Suppressor of variegation 3-9 homolog 2|Su(var)3 9 Drosophila homolog of 2|Histone H3-K9 methyltransferase 2|Histone lysine N methyltransferase SUV39H2|Su(var)3 9 homolog 2|Suv39h2|H3-K9-HMTase 2|Lysine N-methyltransferase 1B|Histone lysine N methyltransferase H3 lysine 9 specific 2|H3 K9 HMTase 2|KMT1B|SUV92_HUMAN|sSuppressor of variegation 3 9 homolog 2 (Drosophila)|Suppressor of variegation 3 9 homolog 2|Su(var)3-9 homolog 2|Histone H3 K9 methyltransferase 2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human KMT1B
- Purification:Antigen affinity purified
- Cat. no.:77699-837
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: SUV39H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse