Specifications
- Description:Fume Extractor
- Cat. no.:CA89087-828
- Supplier No.:70511834
Specifications
About this item
Enables three simultaneous users to capture and filter noxious fumes that occur when soldering, gluing, and working with solvents.
The fan has a variable speed controller that regulates the amount of airflow through the system. Unit is supplied with three extraction arms with an inside diameter of 3.2cm (11/4"). The arms have a 360° swivel at the base and will hold their positions in all directions. Each arm contains a damper to allow for individual users to regulate their own airflow. The clear extraction hood supplied with the kit does not obstruct the view.
The fan has a variable speed controller that regulates the amount of airflow through the system. Unit is supplied with three extraction arms with an inside diameter of 3.2cm (11/4"). The arms have a 360° swivel at the base and will hold their positions in all directions. Each arm contains a damper to allow for individual users to regulate their own airflow. The clear extraction hood supplied with the kit does not obstruct the view.