Dual Exhaust Adapter, Labconco
Catalog #: 76045-692
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Dual Exhaust Adapter
- Cat. no.:76045-692
Specifications
About this item
This accessory for Protector® ClassMate® Fume Hoods is a powder-coated steel adapter
It allows two Protector® ClassMate® Hoods mounted back-to-back to be exhausted from a single 30.5 cm (12") nominal diameter duct. Connects (2) 12" exhaust collar connections to one 12" connection.
Customers Who Bought This Also Bought