About this item
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 plates per meter for the 5 µm particle size, Purospher™ STAR columns ensure good baseline separations. Special derivatization and endcapping processes ensure long-term chemical stability, excellent retention stability, and long column lifetime. All Purospher™ STAR columns are guaranteed to provide absolutely reproducible results.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA1.50624.0001
- CA1.50625.0001
- CA1.50623.0001
- CA1.50626.0001
- CA1.50627.0001
- CA48219-486
- CA48219-488
- CA48219-482
- CA48219-484
- CA48219-825
- CA48219-821
- CA48219-817
- CA48219-812
- CA48219-810
- CA48219-870
- EM1.50750.0001
- CA48219-760
- CA48219-754
- CA1.50256.0001
- CA1.51918.0001
- CA48219-772
- CA48219-770
- CA48219-768
- CA48219-766
- CA48219-789
- CA48219-787
- CA48219-784
- CA48219-782
- CA48219-480
- CA48219-800
- CA48219-798
- CA48219-796
- CA48219-476
- CA48219-478
- CA48219-794
- CA48219-472
- CA48219-793
- CA48219-790
- CA48219-474
- CA1.50254.0001
- CA48219-470
- CA1.50253.0001
- CA48219-808
- CA48219-806
- CA48219-805
- CA48219-803
- Description:
- Purospher® STAR RP-18 Endcapped HPLC Cartridge
- Purospher® STAR RP-18 Endcapped HPLC Cartridge
- Purospher® STAR RP-18 Endcapped HPLC Cartridge
- Purospher® STAR RP-18 Endcapped HPLC Cartridge
- Purospher® STAR RP-18 Endcapped HPLC Cartridge
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR NH2 LiChroCART® Guard Column
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR RP-8 endcapped (5µm) LiChroCART® 125-3 HPLC cartridge
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR RP-18 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-18 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 75-4 HPLC cartridge
- Purospher® STAR RP-8 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 55-4 HPLC cartridge
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 30-4 HPLC cartridge
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR Phenyl Hibar® RT HPLC column
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 55-4 Cartridge set 1 HPLC cartridge and 1 cartridge holder
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 55-2 HPLC cartridge
- Purospher® STAR RP-18 endcapped (3 µm) LiChroCART® 55-2 Cartridge set 1 HPLC-cartridge and 1 cartridge holder
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR SI LiChroCART® HPLC cartridge
- Purospher® STAR SI LiChroCART® HPLC cartridge
- Purospher® STAR SI LiChroCART® HPLC cartridge
- Purospher® STAR SI LiChroCART® Guard Column
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR RP-8 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-8 endcapped (5µm) LiChroCART® 250-4.6 HPLC cartridge
- Purospher® STAR RP-8 endcapped (5µm) LiChroCART® 150-4.6 HPLC cartridge
- Purospher® STAR NH2 LiChroCART® HPLC cartridge
- Guard Column
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR NH2 LiChroCART® HPLC cartridge
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Guard Column
- Purospher® STAR NH2 LiChroCART® HPLC cartridge
- LiChroCART® Stainless Steel Purospher™ STAR Cartridges
- Purospher® STAR NH2 LiChroCART® HPLC cartridge
- LiChroCART® Stainless Steel Purospher® STAR Cartridges
- Purospher® STAR RP-18 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-8 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-8 endcapped Hibar® RT HPLC column
- Purospher® STAR RP-8 endcapped Hibar® RT HPLC column
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5
- 1.5 to 10.5