Snap-n-Slide™ Wall Shelves, Eagle MHC®
Supplier: Eagle Group
Certificates
About this item
Heavy gauge stainless steel shelving system allows positioning of shelves anywhere on the wall brackets without the use of tools or fasteners
- Shelf brackets can be mounted to wall studs
- No wall backing required
Shelves feature a 38mm (1¹/₂") rolled front edge, except for Type 316, 38.1cm (15") wide shelves that feature a 45mm (1³/₄") V-shaped marine edge. Both shelf sizes have a 38mm (1¹/₂") upturn on rear and ends.
Certifications: NSF approved.
Ordering information: Supplied with stainless steel wall brackets. The 61cm (24") and 91.5cm (36") long shelves are supplied with two brackets; the 121.9cm (48") and 152.4cm (60") long shelves are supplied with three brackets; the 182.9cm (72") long shelves are supplied with four brackets.
Customers Who Bought This Also Bought