Specifications
- Height:78½ - 86½"
- W×D:48×30"
- Supplier No.:GL105
Specifications
About this item
All stainless steel workstation is equipped with an electro-mechanical actuator that can raise or lower the work surface from 85.1 to 105.4 cm (33¹/₂ to 41¹/₂")
- allow hands-free operation for hot water, cold water, rinse, and the ¹/₂ hp commercial-grade disposal unit. Lighting is provided by a top-mounted
The height adjustment is stabilized by an internal scissor lift and a rear stabilizer track. Workstation features a large integral sink with mixing faucet and a spray hose assembly with a hand control. Sink basin measures 40.6W×30.5D×25.4H cm (16×12×10"). A dissecting area rinse provides a constant flow of water to work area. The small specimen rinse has a cold water control valve and a sink rinse basket with a stainless steel holder. Infrared controls. fluorescent light fixture and one magnifying lamp with a 3× magnifier and an incandescent bulb mounted on a flexible arm. The workstation features an exhaust duct for connecting to an existing ventilation system, and is equipped with a formalin container with a spigot, a stainless steel paper towel holder, a magnetic instrument holder, a polyethylene dissecting board, and waterproof GFCI receptacles.
Electrical: 115 V, 60 Hz, 20 A disposal, 115 V, 60 Hz, 15 A station.