ContactUsLinkComponent
Laboratory Pumps, Oil-Less Reciprocating, Gast®
Laboratory Pumps, Oil-Less Reciprocating, Gast®
Catalog #: 54907-086
Supplier:  Gast Manufacturing
Oil-less Reciprocating Laboratory Pump
Laboratory Pumps, Oil-Less Reciprocating, Gast®
Catalog #: 54907-086
Supplier:  Gast Manufacturing
Specifications
  • Description:
    Oil-Less Reciprocating Pump
  • Cat. no.:
    54907-086
  • Supplier No.:
    1HAB25M100X
Specifications
About this item
Special purpose unit functions as both vacuum pump and compressor
Features include oil-less operation, low maintenance and rugged construction. The motor is a single phase, 1/6 hp that delivers 1725rpm.
Ordering information: Complete with gauges, hose barbs, regulators, cord with on/off switch, and carrying handle.
Customers Who Bought This Also Bought