ContactUsLinkComponent
VWR® Plate Roller
VWR® Plate Roller
Catalog #: 60941-118
Supplier:  VWR International
VWR* PLATE ROLLER, FILM AND FOIL CS1
VWR® Plate Roller
Catalog #: 60941-118
Supplier:  VWR International
Specifications
  • Description:
    VWR® Plate Roller
  • Cat. no.:
    60941-118
Specifications
About this item
This 10.2 cm (4") soft rubber roller is designed for assuring tight and consistent application of adhesive films and foils to microplates.
  • Tight and consistent application
  • Secure seal around all microplate wells
Customers Who Bought This Also Bought