PS-90 SensaTemp® Production Soldering Iron, PACE®
Catalog #: 30138-506
Supplier: Pace
Specifications
- Description:Production Soldering Iron
- Cat. no.:30138-506
- Supplier No.:6993-0199-P1
Specifications
About this item
For heavy-duty soldering and general-purpose and SMT rework operations
Provides high thermal capacity and flexibility. Handpiece features PermaGround™ tip grounding system that ensures component safety.
Ordering information: Comes with tip and tool stand. For soldering tips, see 30138-116.
Customers Who Bought This Also Bought