VWR® White Films for Luminescence and Microscopy
Catalog #: 89087-696
Supplier: VWR International
Specifications
- Optical property:Light blocking
- Sterility:Non sterile
- Description:White vinyl films
- Thickness:240,0 µm
- Cat. no.:89087-696
Specifications
About this item
These films are 240 µm thick white vinyl with adhesive, for use with white microplates.
- Backing split on the long dimension aids in application of film to plates
- Film can be applied to either top or bottom
- No end tabs
- Recommended temperature range −40 to +80 °C
Light reflective properties of the film, when used to seal clear-bottom plates, maximise the sensitivity of bioluminescence and chemiluminescence assays in bottom-reading Luminometers. White films have also been used as a backing matrix for microscopy of punch-out samples from filter plates.
Each film L×D: 116×76 mm (49/16×3").
Customers Who Bought This Also Bought