About this item
Compact, microprocessor-controlled unit stains up to 20 slides or coverslips.
Optional circulating water bath 25445-014 provides clean rinse water.
- Input up to nine user-programmable procedures of 29 steps each
- Each step may be timed from 1 second to 99 minutes, 59 seconds
- Optional circulating water bath 25445-014 provides clean rinse water
Compact, microprocessor-controlled unit stains up to 20 slides or coverslips.
Unit contains four staining/reagent stations, one running water rinse station programmed to maintain 300mL increments, allowing for precision rinsing, and one forced-air drying station. The rinse and drying stations are easily converted for staining, permitting a maximum of six staining/reagent stations.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA64000SM-94
- CA15204-300
- CA10183-816
- CA6405294
- CAD64054W-95
- CAD64054W95
- CA64052-94
- Overall dimensions:
- 53L x 45W x 38H cm (21 x 18 x 15")
- Electrical:
- 120/230V, 50/60Hz, 5A
- Unit weight:
- 13 kg (28 lbs.)
- Shipping weight:
- 27 kg (60 lbs.)