2400 Disposable N95 Particulate Respirators, Moldex®
Catalog #: 56219-026
Supplier: Moldex/Metric
Small Business Enterprise
Specifications
- Description:2400 N95 Particulate Respirator
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:56219-026
- Supplier No.:2400N95
Specifications
About this item
Designed for use against nuisance levels (less than OSHA Permissible Exposure Limit) of organic vapor odors and/or ozone
- Provides a filter efficiency level of 95% or greater against particulate aerosols free of oil
- Suitable for most welding fumes, and fits easily under welding hoods
- Flame-retardants added to the Dura-Mesh® shell
- Workers stay cool with an exhale valve that reduces breathing resistance and hot air build-up
- Facepiece is naturally contoured without pressure points
- Dura-Mesh® shell resists collapsing, even in heat and humidity
Workers stay cool with an exhale valve that reduces breathing resistance and hot air build-up. Facepiece is naturally contoured without pressure points. Dura-Mesh® shell resists collapsing, even in heat and humidity. Stays cleaner-looking longer.
Suitable for most welding fumes. Fits easily under welding hoods. Flame-retardants added to the Dura-Mesh® shell. (Note: user is responsible for checking current OSHA regulations and applicable guidelines for welding applications.)
Customers Who Bought This Also Bought