ContactUsLinkComponent
8511 N95 Respirator, 3M™
  CA56621-936
 :  3M
undefined
8511 N95 Respirator, 3M™
  CA56621-936
 :  3M
 :  70070757557
  • Description:
    8511 N95 Respirator
  • Environmentally Preferable:
  • Cat. no.:
    CA56621-936
  • Supplier No.:
    70070757557
Respirator provides protection against certain solid and liquid particles that do not contain oil.
  • Provides comfortable protection against certain non-oil based particles
  • Cool Flow™ exhalation valve reduces heat and humidity buildup
  • Features braided headbands, soft inner materials, adjustable "M" nose clip
The innovative “M” nose clip helps provide a custom fit and secure seal against the face. Comfortable, durable respirator has the Cool Flow® exhalation valve to reduce heat and humidity build-up inside the respirator. Provides minimum filtration efficiency of 95%.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.