Barnstead/Lab-Line Unwire™ Test Tube/Microcentrifuge Tube Racks for Universal Platforms, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for use with universal platforms (see 57018-990 series)
ResMer® resin construction withstands extreme conditions, including autoclaving, mechanical washing, chemical disinfection, freezing, and temperatures up to 135 °C (275 °F). Racks are chemically resistant to low concentrations of acids and hydrocarbons, bases, alcohols, esters, and aldehydes.
Ordering information: Racks include a removable polyethylene foam insert to hold tubes tightly and prevent spinning.