Anti-Theft Devices, METTLER TOLEDO®
Supplier: Mettler Toledo
Certificates
About this item
Special locking tubes, plates and pins help keep your balances in place.
- Keep balances in place
- Secure with standard padlock
Heavy aluminum rods fit through special holes in the balances and through a 1.6 cm (5/8") hole drilled through a laboratory benchtop or table. A sleeve or pin fits through the rod and secures with a standard padlock. Series B, BD, PR, SR, PG-S, SG, AB-S, PB-S, and SB balances can use a cable and lock with key.
BodyTextAccessories: Locks are included with device.
Customers Who Bought This Also Bought