Inlet Filter, Labconco®
Supplier: Labconco
Certificates
About this item
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
Delivery: Includes filter assembly and one filter cartridge.