About this item
The illustra GFX PCR DNA and Gel Band Purification kits are for the isolation and concentration of DNA fragments from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.
- Fast and easy-to-use method with less than 10 min hands-on time
- Flexible 10 to 50 μl elution volume for different DNA concentration needs
- Improved column design with increased maximum volume
- Holding of up to 800 μl liquid volume (950 μl without a cap)
- Two choices of elution buffer provided to fit downstream applications
- Color-coded caps for ease of use
The illustra GFX PCR DNA and Gel Band Purification kits are for the isolation and concentration of DNA fragments from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.
illustra GFX PCR DNA and Gel band purification kit is designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg. The kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin column for the purification of DNA from both solution and agarose gel. Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution. DNA purity is exceptional, 99.5% of contaminants are removed.
Highly pure DNA is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning. Addition of a visual color indicator to the capture buffer ensures optimal pH for maximum DNA binding and recovery.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number28903470_HAND BOOK.pdf 28903470_INSTRUCTIONS 1.pdf 28903470_INSTRUCTIONS 2.pdf 28903470_INSTRUCTIONS 3.pdf 28903470_INSTRUCTIONS.pdf
Manual
95026-730_16959251
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Next-Generation Sequencing (NGS) Solutions