VWR® Versatile 11 mm Dual Crimp-Top/Snap-Cap Vials
Supplier: VWR International
Certificates
About this item
Vials feature a versatile neck that allows closure by either snap-cap seals or aluminum crimp seals.
Vials have 40% wider openings than standard vials to reduce risk of needle damage. Manufactured from clear or amber borosilicate glass or polypropylene.
Ordering information: Vials do not come with caps. Caps are available separately.
Delivery: Convenience kits include 2 ml vials and unassembled snap caps.