ContactUsLinkComponent
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent, Thermo Scientific
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent, Thermo Scientific
Catalog #: CAPI23238
Supplier:  Thermo Scientific
CAS Number:  
Coomassie Plus (Bradford) Assay Reagent
Pierce™ Coomassie Plus (Bradford) Protein Assay Reagent, Thermo Scientific
Catalog #: CAPI23238
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Coomassie Plus (Bradford) Reagent
  • Size:
    300 mL
  • Cat. no.:
    CAPI23238
  • No. of Assays/Kit:
    200 standard assays or 1000 microplate assays
  • Supplier No.:
    23238
Specifications
About this item
The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration
  • Sensitive, fast and consistent protein assay kit
  • Determine protein concentrations from 1 to 1500 μg/mL
  • Convenient microplate or cuvette format
  • Compatible with reducing sugars, reducing substances and thiols
This modification of the well-known Bradford method greatly reduces the tendency of Coomassie reagents to give non linear response curves by a formulation that substantially improves linearity for a defined range of protein concentration. In addition, Coomassie Plus Reagent results in significantly less protein-to-protein variation than is observed with other Bradford-type Coomassie formulations.
The Pierce™ Coomassie Plus Protein Assay is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent to quickly measure (A595 nm) total protein concentration compared to a protein standard.
Customers Who Bought This Also Bought