Remote Control Hot Plates, Type RC2200, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for remote control operation in fume hoods or other corrosive environments.
Control can be placed up to five feet from hot plate for operating safety and convenience
Operating range: 150 to 371°C (300 to 700°F). Percentage input controller maintains temperature stability to ±5°C (±9°F) over the operating temperature range.
Top plate is porcelain-coated steel. Corrosion-resistant, stainless steel body. Control unit is housed in rugged, protective steel case. Line cord from hot plate to control is protected by a special, chemically-resistant cover.
Ordering information: Supplied complete with 1.5m (5') three-wire cord and plug.
Customers Who Bought This Also Bought