VWR® Contour™ Full Height Storage Cabinets
Supplier: VWR International
Certificates
About this item
Standing height steel storage cabinets are 84" high and 123/4", 16", or 22" deep.
- Five adjustable shelves
- Light neutral color
Cabinets are phosphate-coated with a baked, chemical-resistant, powder-coat finish.
Caution: Full height cabinets may present a potential tipping over problem. Proper installation requires these cabinets to be anchored to a wall or other cabinets to improve stability and ensure smooth door movement.
Customers Who Bought This Also Bought