Sink Tops, Just Manufacturing
Supplier: Avantor
Certificates
About this item
Fabricated of Type 304, 18-8 stainless steel with reinforced drainboards
Drainboards are double-pitched to bowl to provide drainage without creases or grooves. Entire undersurface is thoroughly sound deadened. Deck punched with four faucet holes; bowl center-punched for cup strainer.
Top is formed from one sheet of stainless steel and the bowl is seamlessly welded to the drainboard to provide anti-splash rim around entire perimeter of bowl.
Delivery: Provided with wood frames to facilitate mounting.
Customers Who Bought This Also Bought