VWR® Atlas Overhead Storage Cabinets with Sliding Doors
Supplier: VWR International
CE Compliant
Certificates
About this item
Steel cabinets are used in place of open shelving when enclosed storage and/or resistance to dust is required.
- Sliding doors
- Color: Light neutral
For double-sided storage without pass-through, two cabinets of the same size may be mounted back to back.
For use with VWR® Atlas mobile carts/mobile laboratory benches.
Ordering information: Supplied with mounting hardware. Hardware for back-to-back installation not included.
Customers Who Bought This Also Bought