About this item
Self-contained membrane nitrogen generator produces LC/MS grade nitrogen with a maximum output pressure of 116psig and maximum flow capacity of 30L/min.
- Continuous or on-demand nitrogen supply
- Reliable, durable generator
- Special sound insulation design ensures quiet operation
The membrane will not suppress corona needle discharge, unlike PSA and Hosmer technologies
Generator can deliver a continuous or on-demand supply of pure, phthalate-free nitrogen, making it an excellent alternative to cylinders or dewars. Reliable, durable generator has been engineered with oil-free compressors, carefully matched filtration, and membrane separation and compression technologies.
Ambient air is filtered using an inlet suction breather filter. The purified air is delivered to a long-life low pressure air compressor, which provides an air stream to hollow fiber membranes. These membranes separate the clean air into a concentrated nitrogen retentate and oxygen-enriched permeate. Prior to exiting the system, pure nitrogen retentate is delivered to a nitrogen amplification compressor to assure proper pressure, flow, and purity to the LC/MS system.
Additional applications include nebulizer gases, chemical and solvent evaporation, instrument supply and purge, evaporative light scattering equipment, and sparging. Also ideal for derivatives of ESi and APCi modes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA62700-326
- Dimensions:
- 70.1H x 31W x 90D cm (271/2 x 121/4 x 357/16")
- Weight:
- 92.5 kg (204 lbs.)