Accessories for VWR® Atlas Shelving
Supplier: VWR International
Certificates
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
$649.44 to $2,496.86
Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
$102.63 to $271.58
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Mix & Go, <i>E. coli</i> Transformation Kit & Buffer Set, Zymo Research
Supplier: Zymo Research
$204.14 to $204.14
Razor Blades: Double Edge Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$146.53